missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Picus™ NxT Electronic Pipettes, 8 Channels
BIOPHARMAGREENER_CHOICE

Product Code. p-4912124 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
0.2 to 10 μL
10 to 300 μL
5 to 120 μL
50 to 1200 μL
Color:
Gray
Orange
Purple
Yellow
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color unitSize
15320635 0.2 to 10 μL Gray Each
15330635 5 to 120 μL Yellow Each
15340635 10 to 300 μL Orange Each
15337774 50 to 1200 μL Purple Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15320635 Supplier Sartorius Supplier No. LH745321

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed to ease user's workload and to protect from repetitive strain injury (RSI)

Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.

  • Password protection for stored programs
  • Very last liquid droplets dispensed by repeated blow-out for maximum recovery
  • Features one-touch operating-button and feather-light electronic tip ejection
  • Includes plate tracker for microwell plate dispensing
  • Electronic piston control and brake for precise pipetting
  • Safe-cone filters to prevent contamination
  • Adjustment wheel for volume setting and navigation
  • Range of pipetting modes for diluting and titrating to serial dispensing
  • Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
  • Certificate of accredited 3-point calibration
  • RFID-enabled to support asset management
  • Volume range: 0.2 μL to 1200 μL
  • Delivered without USB AC adapter (available separately)

Applications:

PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture

Certifications

CE Marked

Warranty

2 year

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Number of Channels 8
Product Line Picus
Type Electronic Pipette
Volume (Metric) 0.2 to 10 μL
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.