Learn More
Thermo Scientific™ 0.1% Formic Acid (v/v) in Acetonitrile, LC-MS Grade
An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
225.00 € - 792.00 €
Specifications
Grade | LC-MS |
---|---|
Item Name | Formic Acid |
Content And Storage | Store in a cool, dry, and well-ventilated area. Protect from direct sunlight. |
Form | Liquid |
Reagent Type | Solvent Mixture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13404279
|
Thermo Scientific™
85175 |
4 x 1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13494269
|
Thermo Scientific™
85174 |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ 0.1% Formic Acid in Acetonitrile is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
This 0.1% Formic Acid in Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography- and spectrometry-grade 0.1% FA in acetonitrile is 0.2–micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection.Highlights:
Validated
– tested more than 20 ways to ensure maximal LC/MS sensitivity
Purified
– processed, filtered and sealed to extend LC/MS column life
Convenient
– packaged to eliminate variability and reduce handling
Specifications
LC-MS | |
Store in a cool, dry, and well-ventilated area. Protect from direct sunlight. | |
Solvent Mixture | |
HPLC, Mass Spectrometry |
Formic Acid | |
Liquid | |
Pierce™ |
For Research Use Only. Not for use in diagnostic procedures.