missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 0.1% Trifluoroacetic Acid (v/v) in Water, LC-MS Grade

Product Code. 13434279 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13434279 4 x 1 L Pack of 4
13414279 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13434279 Supplier Thermo Scientific™ Supplier No. 85173

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

Thermo Scientific™ 0.1% Trifluoroacetic Acid in Water is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

This 0.1% Trifluoroacetic Acid in Water is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography-and spectrometry-grade 0.1% TFA in water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Validated

    – tested more than 20 ways to ensure maximal LC/MS sensitivity

  • Purified

    – processed, filtered and sealed to extend LC/MS column life

  • Convenient

    – packaged to eliminate variability and reduce handling

TRUSTED_SUSTAINABILITY

Specifications

Grade LC-MS
Item Name Trifluoroacetic Acid (TFA)
Quantity 4 x 1 L
Content And Storage Store in a cool, dry, and well-ventilated area. Protect from direct sunlight.
Form Liquid
Reagent Type Solvent Mixture, Ion Pairing Reagent
Product Line Pierce
For Use With HPLC, Mass Spectrometry

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.