missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Firefly Signal Enhancer (100X)

Product Code. 12132250 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12132250 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12132250 Supplier Thermo Scientific™ Supplier No. 16180
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Produce maximum bioluminescence signal intensity and sensitivity with the Pierce Firefly Luciferase Flash and Glow Assays.

The Thermo Scientific™ Pierce Firefly Signal Enhancer is designed to complement the Pierce Firefly Luciferase Flash and Glow Assay and is recommended when maximum sensitivity is desired.

The Pierce Firefly Signal Enhancer is designed to to be used with the Pierce Firefly Luciferase Flash Assay Kit (Part No. 16174, 16175) and Pierce Firefly Luciferase Glow Assay Kit (Part No. 16166, 16167). When used with the flash assay kit, the enhancer provides a five-fold increase in signal. When used with the glow assay kit, the enhancer solution increases the signal strength and duration up to two-fold and also increases stability of the bioluminescent signal.

Highlights:

  • Up to 5-fold increase in Firefly Flash signal intensity
  • Up to 2-fold increase in Firefly Glow signal intensity and duration
  • Convenient and easy-to-use 100X solution

Requires:

Pierce Firefly Luciferase Flash (Part No. 16174, 16175) or Glow (Part No. 16166, 16167) Assay Kits; Luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.

TRUSTED_SUSTAINABILITY

Specifications

Type Firefly Signal Enhancer
Quantity 0.5 mL
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Detection Method Bioluminescence
Substrate Properties Chemical Substrate
Substrate Type Luciferase Substrate
Product Line Pierce

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.