missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 10X Western Blot Transfer Buffer, Methanol-free

Product Code. 12691410 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
5 L
Unit Size:
5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12691410 500 mL 500mL
10565604 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12691410 Supplier Thermo Scientific™ Supplier No. 35045

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A methanol-free concentrated solution to easily and quickly prepare MeOH-free Western blot transfer buffer for tank-based or other wet gel-to-membrane transfer methods.

Thermo Scientific Pierce 10X Western Blot Transfer Buffer, Methanol-free is a space-saving stock solution for preparing the methanol-free transfer buffer used with cooled, tank (wet) Western blotting transfer appratus.

Pierce Concentrated Buffers are ready to use without having to weigh and dissolve dry ingredients or to adjust the pH with concentrated acid or base. Simply dilute the stock solution with pure water and proceed with your experiment. The 10X Western Blot Transfer Buffer is ready to use upon dilution to 1X with water; it does not contain methanol and the addition of methanol is not necessary for use in electrophoresis.

Highlights:

  • Transfer Buffer – diluted 10-fold in water, the solution is ready to use for electrophoresis (i.e., wet tank transfer from mini gels)
  • Easy to use – no packets to open, no powder to dissolve, and no methanol required
  • Increased accuracy – eliminates the possibility of powder remaining in a packet
  • Saves time – 10X concentration eliminates time spent waiting for powder to dissolve
  • Saves space – storage as concentrated stock minimizes bench space needed for solutions

Recommended for:

Wet (tank) Western blot transfer of proteins to a membrane

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage: Upon receipt store at 4°C, protected from moisture. Product is shipped at ambient temperature.
Buffer Transfer Buffer
Concentration 10X
Gel Type SDS-PAGE
Gel Compatibility SDS-PAGE Gels
Quantity 500 mL
Shipping Condition Ambient
Product Line Pierce
Product Type Western Blot Transfer Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.