missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Amino Acid Standard H

Product Code. 13447076 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13447076 10 x 1 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13447076 Supplier Thermo Scientific™ Supplier No. TS20088

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Calibrate HPLC analysis of protein hydrolysates using this calibration standard mixture of 18 amino acids, each at 2.5 micromoles/mL in 0.1N HCl.

Thermo Scientific™ Pierce Amino Acid Standard H is a quantitative mixture of 18 amino acids, supplied at 2.5μmol/mL in 0.1N HCl, for use as a high-purity calibration standard for HPLC analysis of protein hydrolysates.

Highlights:

  • Uses L-form configuration to permit standardization of microbial and other assays
  • Molar concentration verified by conventional amino acid analysis methods
  • With the exception of cystine, each amino acid is supplied at a concentration of 2.5μmoles/mL in 0.1N HCl
  • L-alanine, Ammonia [(NH4)2SO4], L-Arginine, L-Aspartic Acid, L-Cystine, L-Glutamic Acid, Glycine, L-Histidine, L-Isoleucine, L-Leucine, L-Lysine·HCl, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tyrosine, L-Valine

Includes:

  • L-alanine, Ammonia [(NH4)2SO4], L-Arginine, L-Aspartic Acid, L-Cystine, L-Glutamic Acid, Glycine, L-Histidine, L-Isoleucine, L-Leucine, L-Lysine·HCl, L-Methionine, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tyrosine, L-Valine
TRUSTED_SUSTAINABILITY

Specifications

Product Type Amino Acid Standard H
Content And Storage Store between -20°C and 8°C.
Quantity 10 x 1 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.