missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ ANB-NOS Crosslinker

Product Code. 10677924 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10677924 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10677924 Supplier Thermo Scientific™ Supplier No. 21451

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Crosslink amines to anything witht this short, photoreactive crosslinker with NHS-ester and nitrophenyl azide reactive groups; photoactivates with UV light (Amax 320nm).

Thermo Scientific™ Pierce ANB-NOS is a short-arm (7.7 Å) crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide.

Highlights:

  • Reactive groups: NHS ester and nitrophenyl azide (Amax= 320nm)
  • Reactive toward: amino groups and various sites
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-to-Nonselective
Cleavable No
Description Pierce ANB-NOS
Formulation N-5-Azido-2-nitrobenzoyloxysuccinimide; crystalline or powder solid
Membrane Permeable No
Molecular Weight (g/mol) 305.2
PEGylated No
Sufficient For 164mL of typical 1X (1mM) reaction solution, or approx. 25 uses at 2mg reagent per use
Spacer Arm Length 7.7Å
Quantity 50 mg
Solubility Not soluble in water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.