missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Anti-DYKDDDDK Affinity Resin

Product Code. 15895833 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 mL
5 Columns
Unit Size:
1mL
50mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15895833 2 mL 1mL
15815843 10 mL 5mL
15825843 100 mL 50mL
31034559 5 Columns -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 15895833 Supplier Thermo Scientific™ Supplier No. A36801
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Pierce Anti-DYKDDDDK Spin Columns provide a fast, convenient method for purification of DYKDDDDK-tagged proteins expressed in in vitro protein expression systems, bacteria, yeast, and mammalian cells.

Thermo Scientific Pierce Anti-DYKDDDDK Spin Columns provide a fast, convenient method for purification of DYKDDDDK-tagged proteins expressed in in vitro protein expression systems, bacteria, yeast, and mammalian cells. The amino acid sequence DYKDDDDK, commonly known as 'FLAG', is recognized by a high-affinity rat monoclonal antibody (clone L5) that is covalently attached to UltraLink Biosupport, which is a rigid, hydrophilic, highly crosslinked, copolymeric, and porous resin with high coupling capacity.

For protein purification, resin is added to a sample containing DYKDDDDK-tagged proteins with the tag on either the N- or C-terminus. Following capture of DYKDDDDK-tagged proteins, non-specifically bound proteins can be washed away before dissociating bound target proteins with elution buffer.

Features of Pierce Anti-DYKDDDDK Affinity Resin

  • Specific—unique base beads and highly specific antibody minimize off-target binding (low non-specific binding)
  • High purity—optimized bind-wash-elute protocol enables high purity
  • High yield—special antibody conjugation method enables high yield
  • Rapid—entire purification protocol typically takes less than 40 mins
  • Economical—purification protocol allows multiple reuses
  • Flexible—available in multiple formats including bulk loose resin and spin columns

Characteristics of Pierce Anti-DYKDDDDK Affinity Resin

  • Composition: anti-DYKDDDDK antibody covalently attached to UltraLink Biosupport, a highly crosslinked rigid support
  • Particle size: 50–80 μm
  • pH stability: 1–13
  • Operating temperature: 2–30°C; do not freeze
  • Particle concentration: 50% slurry in phosphate buffered saline, 0.02% sodium azide, pH 7.5
  • Binding capacity: ≥3.0 mg DYKDDDDK-tGFP-His protein (∼32 kDa/mL settled resin)

Available formats

  • Resin slurries—2 mL, 10 mL, and 100 mL of 50% slurry in PBS with 0.02% sodium azide, pH 7.5
  • Spin columns—10 mL spin columns containing 3 mL of settled resin, supplied at 6 mL 50% slurry
TRUSTED_SUSTAINABILITY

Specifications

Product Line Pierce
Product Type Affinity Resin
Quantity 2 mL
Formulation 50% suspension in PBS containing 0.02% Sodium azide
Content And Storage 1 mL settled resin. 2 mL supplied as a 50% slurry in PBS containing 0.02% sodium azide. Store at 2–8°C.
Format Liquid
Stationary Phase Anti-DYKDDDDK monoclonal antibodies
Shipping Condition Wet Ice
Form Liquid
Matrix Affinity Resin
What is the slurry/suspension percentage for your anti-DYKDDDDK ("FLAG") products?

Anti-DYKDDDDK Magnetic Agarose (Cat. Nos. A36797, A36798, A36799B) is offered as a 25% suspension (1 mL of 25% suspension = 0.25 mL of settled beads). UltraLink-based Anti-DYKDDDDK Affinity Resin (Cat. Nos, A36801, A36803, A36804) is offered as a 50% slurry (1 mL of 50% slurry = 0.5 mL of settled resin).

What is the binding capacity for your Anti-DYKDDDDH Affinity Resin?

Here is the binding capacity: ≥3.0 mg DYKDDDDK-tGFP-His protein (˜32 kDa)/mL settled resin.

Can I use your UltraLink-based Anti-DYKDDDDH ("FLAG") Affinity Resin for FPLC purification?

Yes. The UltraLink-based Anti-DYKDDDDH (“FLAG”) Affinity Resin (Cat. Nos. A36801, A36803, A36804) can be used for FPLC purification.

How do I cleave off the DYKDDDDK ("FLAG") tag after purification?

An enterokinase cleavage site behind the DYKDDDDK (“FLAG”) tag can allow complete removal of the DYKDDDDK (“FLAG”) tag leaving no additional amino acids. We offer EKMax Enterokinase (Cat. Nos. E18001 and E18002) that can be used for this purpose. Subsequently, the EKMax Enterokinase can be removed using EK-Away Resin (Cat. No. R18001), a resin conjugated with soybean trypsin inhibitor, which has high affinity for enterokinase.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.