missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Aquaporin Z Membrane Protein Standard

Product Code. 30089626 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 μg
5 x 20 μg
Unit Size:
20µg
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30089626 20 μg 20µg
30088670 5 x 20 μg Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089626 Supplier Thermo Scientific™ Supplier No. A66693
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Thermo Scientific™ Pierce Aquaporin Z Membrane Protein Standard is a highly validated pure membrane protein formulated for direct infusion native mass spectrometry experiments.

The Thermo Scientific™ Pierce™ Aquaporin Z Membrane Protein Standard is a highly validated pure membrane protein formulated for direct infusion native mass spectrometry experiments.

Features of Pierce Aquaporin Z Membrane Protein Standard include:

  • Positive control sample – aquaporin Z protein production optimized and validated for use in native mass spectrometry
  • Rigorously tested – high quality, consistent protein standard documented via lot-specific certificate of analysis
  • Convenient – ready-to-use format with each single-use vial sufficient for 5 injections

Achieve confidence in your methods without wasting precious samples
Method development for membrane protein analysis provides unique challenges. Pierce Aquaporin Z Membrane Protein Standard empowers success by letting you test parameters and troubleshoot issues without wasting irreplaceable samples. This high purity standard contains sufficient material for multiple qualitative experiments by direct infusion or LC/MS. You can use it to verify system performance, tune parameters, and develop native LC/MS methods without worrying about depleting your real samples.

TRUSTED_SUSTAINABILITY

Specifications

Description Tetramer
Formulation Liquid in 0.05% LDAO, 200 mM Ammonium Acetate; pH 7.4
For Use With (Equipment) Q-Exactive™ UHMR, HMRn option on Tribrid Series MS systems
Material Aquaporin Z
Product Type Protein Standard
Quantity 20 μg
Color Colorless
Grade Mass Spec Grade
Molecular Weight (g/mol) 98,120 Da
Form Liquid

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.