missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BMPH (N-β-maleimidopropionic acid hydrazide)

Product Code. 10157633 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 10157633

Brand: Thermo Scientific™ 22297

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this short (8.1A) crosslinker containing maleimide and hydrazide groups.

Thermo Scientific™ Pierce BMPH is a short, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).

Highlights:

  • Water-soluble
  • Reactive groups: maleimide and hydrazide
  • Reactive toward: sulfhydryl groups and carbonyl aldehyde groups (or carboxyl groups when used with EDC)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Carbohydrate-Sulfhydryl
Cleavable No
Description BMPH
Molecular Weight (g/mol) 297.19
PEGylated No
Spacer Arm Length 8.1 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Amine, Hydrazide, Maleimide
Spacer Short (<10 Å)
Form Solid
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.