missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ BS2G-d0 and BS2G-d4 Crosslinker Mass Pair

Product Code. 10511104 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10511104 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10511104 Supplier Thermo Scientific™ Supplier No. 21615

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Crosslink amines to amines using these heavy (deuterated) and light matched pairs of a 7.7A, NHS-ester crosslinker for mass spectrometry analysis of protein interactions.

Thermo Scientific™ Pierce BS2G-d0/d4 reagents form a matched set of heavy (deuterated) and light mass varieties of a short, amine-reactive crosslinker that facilitates the identification of crosslinked peptides by mass spectrometry.

Highlights:

  • Well-characterized, high-purity, deuterium-labeled crosslinker and its hydrogen-containing analog
  • Instructions edited by an expert in the crosslinking/MS strategy field
  • Suberate- and glutarate-based reagent pair - provides a “molecular ruler” option to study inter- and intra-molecular distances to gain structural information not possible with just one reagent
  • Efficient and convenient - requires only microgram amounts of protein; an excellent alternative to NMR or X-ray crystallography-based methods that require large amounts of protein, special solvents or crystal formation
  • Reactive groups: Sulfo-NHS ester (homobifunctional)
  • Reactive toward: amino groups
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce BS2G-d4
Formulation Bis(sulfosuccinimidyl) 2,2,4,4-glutarate-d4; powder solid
Sufficient For 19mL of typical 1X (1mM) reaction solution, or approx. 5 uses at 2mg reagent per use
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.