missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Cell Surface Biotinylation and Isolation Kit

Product Code. 16462085 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8 Samples
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462085 8 Samples Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16462085 Supplier Thermo Scientific™ Supplier No. A44390
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Thermo Scientific Pierce Cell Surface Protein Biotinylation and Isolation Kit enables the selective biotinylation, solubilization, and enrichment of plasma membrane proteins.

Features of the Cell Surface Protein Biotinylation and Isolation Kit include:
Improved performance—kit and protocol have been re-designed to improve enrichment and extraction of biotinylated plasma membrane proteins while minimizing intracellular protein contamination
Optimized—protocol has been refreshed to reduce processing time and the number of steps, and reagent formulations optimized for improved performance
Compatible—reagents have been verified with suspension and adherent cell lines, with additional instructions provided for MS sample preparation
Validated applications—reagents and procedures are compatible with western blotting and MS applications

This easy-to-use kit provides all of the necessary components and procedure for optimal labeling and subsequent isolation of cell surface proteins. Buffers are supplied pre-formulated to produce consistent results. Mammalian cells are first labeled with EZ-Link Sulfo-NHS-SS-Biotin, a thiol-cleavable amine-reactive biotinylation reagent. Cells are subsequently lysed and the labeled proteins are captured with NeutrAvidin Agarose. Dithiothreitol (DTT) is used in the elution to reduce the disulfide bonds in the biotin label, resulting in the release of the bound proteins without the biotin label. This kit contains sufficient reagents for eight experiments, consisting of two 85-95% confluent 15-cm dishes or four 75-cm2 flasks.

Cell surface proteins represent a key subset of cellular proteins and play major roles in signal transduction, cell adhesion, and ion transport. These proteins are challenging to efficiently extract and isolate due to their multiple membrane-spanning domains. Often, the harsh detergents and conditions necessary for efficient extraction result in denaturation and contamination of plasma membrane proteins after isolation. As an alternative, this kit utilizes a membrane-impermeable, amine-reactive, sulfo-NHS-SS-biotin to efficiently label cell surface proteins with accessible lysines. The lysis, capture, wash, and elution conditions have been optimized for efficient enrichment and isolation while minimizing contamination of intracellular proteins.

The protocol has been optimized for use with diverse mammalian cell lines, including adherent and suspension cells, and is useful for differential expression analysis between treated and non-treated cells or between one or more cell lines. Selectivity and efficiency of the cell surface protein isolation has been verified by western blotting and LC-MS analysis.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • EZ-Link Sulfo-NHS-SS-Biotin, 8 x 6 mg vials, store at 2°C to 8°C
• Lysis Buffer, 10 mL, store at 2°C to 8°C
• NeutrAvidin Agarose, store at 2°C to 8°C
• Wash Buffer, 35 mL, store at 2°C to 8°C
• Elution Buffer, 5 mL, store at 2°C to 8°C
• Column Accessory Pk., 16 spin columns, 16 caps, 16 bottom plugs, and 16 collection tubes, store at room temperature
• Dithiothreitol (DTT), No-Weigh Format, 1 x 7.7 mg microtube, store at 2°C to 8°C
• BupH Phosphate Buffered Saline Pk., 2 Pk., store (at room temperature
• BupH Tris Buffered Saline Pk., 2 Pk., RT)
Shipping Condition Room Temperature
Product Line Pierce
Quantity 8 Samples
Product Type Isolation Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.