missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Compat-Able™ Protein Assay Preparation Reagent Kit

Product Code. 11861345 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11861345 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11861345 Supplier Thermo Scientific™ Supplier No. 23215
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Precipitation reagents to eliminate interfering substances from protein assay samples to make them compatible with BCA and Coomassie-Bradford assays.

Thermo Scientific™ Compat-Able Protein Assay Preparation Set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.

The Compat-Able Reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods. The procedure is accomplished in less than 10 minutes with just a few simple steps. The Compat-Able Preparation Set is available individually or conveniently bundled with either the Thermo Scientific Pierce BCA Protein Assay Kit or the Thermo Scientific Pierce Coomassie Plus Protein Assay Kit.

Highlights:

  • Eliminate incompatibilities – removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer

  • Simple and fast – prepare samples in less than 10 minutes with four simple steps

  • Convenient – reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results

  • Scalable – microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs

  • Validated – tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalization procedures

Includes:

Set contains two bottled reagents (Reagent 1 and Reagent 2); Complete kits contain the Reagent Set plus components of the respective protein assay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Kit contains sufficient reagents to pretreat up to 500 samples
• 250 mL Compat-Able Protein Assay Preparation Reagent 1
• 250 mL Compat-Able Protein Assay Preparation Reagent 2

Store at room temperature
Description Compat-Able Protein Assay Preparation Reagent Set
Assay BCA or Bradford Assay
Detection Method Colorimetric
Product Line Compat-Able
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
For Use With (Application) Incompatibility Elimination
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 500 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Compat-Able Protein Assay Preparation Reagent Kit
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305-IF IN EYES:.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P311-Call a POISON CENTER/doctor/ .
  • P331-Do NOT induce vomiting.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Trichloroacetic acid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.