missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Crossflow Diafiltration Reservoir, 500 mL
Shop All Thermo Scientific ProductsDescription
The Pierce Crossflow Diafiltration Reservoir allows for continuous buffer replacement using Pierce Cross Flow Filtration Cassettes.
• For continuous buffer replacement using Pierce Cross Flow Filtration Cassettes.
Specifications
Specifications
| Color | Clear |
| Material | Plastic |
| Sterility | Non-sterile |
| Type | Self-standing Reagent Reservoir with Lid |
| Includes | 3 ports |
| Quantity | Each |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| Bottom Shape | Flat |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?