missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Crosslink Magnetic IP/Co-IP Kit

Product Code. 11336012 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
40 reactions kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11336012 40 reactions kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11336012 Supplier Thermo Scientific™ Supplier No. 88805
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Immunoprecipitate target proteins without antibody contamination by crosslinking IP antibodies to Protein A/G magnetic beads.

The Thermo Scientific Pierce Crosslink Magnetic IP/Co-IP Kit uses crosslinking chemistry to covalently immobilize IP antibodies onto premium-quality Protein A/G Magnetic Beads for effective immunoprecipitation and co-immunoprecipitation.

Because this magnetic IP procedure involves covalent attachment of the specific antibody used for immunoprecipitation, target proteins or co-IP protein complexes can be eluted and analyzed without antibody fragments because the antibody remains affixed to the beads. The kit contains an optimized protocol and all buffers and reagents necessary to accomplish high-yield IP or co-IP with antibodies that bind to Protein A/G and using either manual or automated magnetic separation tools.

Features of the Crosslink Magnetic IP/Co-IP Kit:

  • Antibody-free IP—antibody is irreversibly attached to the magnetic beads resulting in negligible co-elution of intact antibody or heavy and light chains with the purified antigen
  • Convenient—the IP/co-IP kit contains magnetic beads and all essential buffers for optimal immunoprecipitation
  • Fast—complete crosslinking and immunoprecipitation in less than 3 hours
  • Efficient—immunoprecipitate with half the recommended volume of magnetic particles compared to other magnetic beads
  • Compatible—magnetic beads coupled with Protein A/G recombinant protein ensures compatibility with most primary antibodies, whether from mouse or rabbit
  • Scalable—use only the amount of antibody needed for a single IP experiment or prepare a larger quantity of antibody-crosslinked magnetic beads for multiple experiments

The protocol for the Pierce Crosslink Magnetic IP/Co-IP Kit binds IP antibody to Protein A/G Magnetic Beads (Catalog #88802) and then covalently crosslinks the antibody to the beads using disuccinmidyl suberate (DSS) (Catalog #21655). The antibody-crosslinked beads are then incubated with cell lysate or tissue extract that contains the protein antigen of interest, allowing the antigen:antibody complex to form. The beads are washed to remove non-bound material and a low pH elution buffer is used to dissociate bound antigen from the antibody-crosslinked beads. Neutralization buffer is included to prevent precipitation of the isolated antigen and to ensure protein activity in downstream applications. Lane Marker Sample Buffer is included for preparing samples for SDS-PAGE analysis. The protocol is optimized for 2 to 10μg of IP antibody. For optimal co-IP yields, use 5 to 10μg of antibody. The beads are removed from the solution manually using a magnetic stand or by automation with an instrument such as the Thermo Scientific KingFisher Flex Instrument.

Traditional IP (without covalent antibody attachment) generally provides higher antigen yields than IP protocols that used crosslinking. This is because the crosslinking process inevitably causes loss of some functional antibody binding sites. To overcome this limitation with the crosslink method, it may be necessary to use somewhat greater amounts of IP antibody in the crosslink IP method than in the equivalent traditional IP procedure. Of course, the advantage with the crosslink immunoprecipitation procedure is Western blots devoid of interfering antibody bands.

Protocol Summary

  1. Prewash beads with 1X Coupling Buffer.
  2. Bind antibody to Protein A/G magnetic beads for 15 minutes.
  3. Wash beads three times with 1X Coupling Buffer.
  4. Crosslink antibody to beads with DSS for 30 minutes.
  5. Wash bead three times with Elution Buffer and then twice with IP Lysis/Wash Buffer.
  6. Incubate cell lysate with prepared beads for 1-2 hours at room temperature or overnight at 4°C.
  7. Wash beads twice with IP Lysis/Wash Buffer and then once with purified water.
  8. Elute bound antigen.

Related Products

Pierce™ Crosslink IP Kit (Catalog #26147)

TRUSTED_SUSTAINABILITY

Specifications

Formulation Slurry: 10 mg/mL, 1% solids
Content And Storage Sufficient For: 40 IP reactions using 25 μL of beads
• Pierce Protein A/G Magnetic Beads, 1 mL
• IP Lysis/Wash Buffer, 2 x 50 mL
• Coupling Buffer (20X), 25 mL
• DSS Crosslinker, 8 x 2 mg
• Lane Marker Sample Buffer (5X), 5 mL
• Elution Buffer, 25 mL
• Neutralization Buffer, 1 mL
• Lane Marker Sample Buffer (5X), 5 mL

Store at 4°C. Store DSS desiccated at 4°C.
Assay Affinity Purification
For Use With (Equipment) KingFisher™ Flex Magnetic Particle Processor, Magnetic Stand
Product Line Pierce
Quantity 40 reactions kit
For Use With (Application) Immunoprecipitation (IP)
Target Not Target-Specific
Type Immunoprecipitation/Co-Immunoprecipitation Kit
Sufficient For 40 Reactions
Format Kit
Capacity (Metric) 1 mL
Show More Show Less
compliance-icons
Product Identifier
  • Pierce Crosslink Magnetic IP/Co-IP Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P361-Take off immediately all contaminated clothing.
  • P363-Wash contaminated clothing before reuse.
Supplemental information
  • MIXTURE LIST-Contain: sodium chloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.