missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ D-Luciferin, Monosodium Salt, 100 mg

Product Code. 15285723 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15285723 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15285723 Supplier Thermo Scientific™ Supplier No. 88291
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Get convenience and high performance at a low cost in firefly luciferase reporter assays with our D-Luciferin, formulated at greater than 99% purity as both monosodium and monopotassium salts.

Thermo Scientific™ Pierce D-Luciferin is synthesized and manufactured to ensure high performance at a low cost, and the reporter is also tested by HPLC to ensure proper chirality and activity.

We are the only supplier to perform chiral-high performance liquid chromatography (HPLC) testing to ensure that the product is not only pure but also in the active form. Our D-Luciferin outshines the competition on many levels.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -20°C.
Detection Method Bioluminescence
For Use With (Application) ATP Assay, Gene Reporter Assay
Product Type D-Luciferin, Monosodium Salt
Product Line Pierce
Quantity 100 mg
Salt Form Monosodium Salt
Substrate Properties Chemical Substrate
Substrate Type Luciferase Substrate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.