missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNA Coating Solution

Product Code. 10405495 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10405495 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10405495 Supplier Thermo Scientific™ missing translation for 'supplierNo' 17250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Efficiently immobilize DNA to microplate wells and microsample tubes with this DNAse-free coating solution.

Perform efficient DNA surface coating of microplates and microsample tubes using Thermo Scientific™ Pierce DNA Coating Solution.

This simple solution provides the appropriate conditions to promote passive adsorption of purified DNA onto polystyrene surfaces, such as those in standard ELISA-type microplates. DNA becomes coated noncovalently along its length, allowing its detection with certain hybridization methods. Particular probing techniques must be validated empirically to determine compatibility.

Highlights:

  • No more overnight incubations or heating microplates and waiting
  • Promotes efficient immobilization (by adsorption) of DNA in microplate wells (polystyrene)
  • Coated target DNA permits speed and flexibility in probing by hybridization and other means
  • DNA coated onto plastic surfaces is stabilized sufficiently to allow storage for months before use
  • Solution has been autoclaved to inactivate DNase
TRUSTED_SUSTAINABILITY

Especificaciones

Buffer Immunoassay Buffer, Hybridization Buffer
Content And Storage Store at room temperature.
Description Pierce DNA Coating Solution
Formulation Proprietary buffered solution
Product Type DNA Coating Solution
Quantity 100 mL
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.