missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DTBP (Wang and Richard's Reagent)

Product Code. 10762725 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10762725 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10762725 Supplier Thermo Scientific™ Supplier No. 20665
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Crosslink amines to amines with dithiobispropionimidate; composed of imidoester groups at either end of a reducible (cleavable) disulfide-containing spacer arm.

Thermo Scientific™ Pierce DTBP is dimethyl 3,3'-dithiobispropionimidate, a crosslinker that contains amine-reactive imidoester groups around an 8-atom spacer arm, whose central disulfide bond can be cleaved with reducing agents.

Highlights:

  • Water-soluble
  • Cleavable by reducing agents
  • Penetrates intact human erythrocytes
  • Reactive groups: imidoester (homobifunctional)
  • Reactive toward: amino groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable By Thiols
Description DTBP
Molecular Weight (g/mol) 309.28
PEGylated No
Spacer Arm Length 11.9 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Imidoester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 1 g
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
compliance-icons
Product Identifier
  • DTBP (Wang and Richard's Reagent)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Dimethyl 3,3'-Dithiobispropionimidate 2HC

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.