missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EDTA

Product Code. 11836714 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11836714 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11836714 Supplier Thermo Scientific™ Supplier No. 17892
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chelate alkaline earth metals. Thermo Scientific™ Pierce EDTA is highly purified for use in a variety of laboratory methods.

Thermo Scientific™ Pierce EDTA is useful as a chelator of alkaline earth metals, as well as iron, copper and zinc in a variety of laboratory methods.

Additional Information and Online Ordering

EDTA (ethylenediaminetetraacetic acid) is useful as a chelator of alkaline earth metals, as well as iron, copper and zinc. EDTA is especially useful for its ability to sequester metal ions to reduce their reactivity in solutions.

  • White crystalline powder
  • ≥99.0% Pure
  • Water soluble
  • MW: 372.24 g/mol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EDTA
CAS 6381-92-6
Content And Storage Store at room temperature.
Molecular Formula C10H14N2Na2O8·2H2O
Product Type EDTA (ethylenediaminetetraacetic acid)
Purity ≥99%
Quantity 500 g
Synonym EDTA disodium salt dihydrate
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.