missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EMCS (N-ε-malemidocaproyl-oxysuccinimide ester)

Product Code. 10147783 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 10147783

Brand: Thermo Scientific™ 22308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Crosslink amines to sulfhydryls with this mid-length crosslinker composed of NHS-ester and maleimide groups at ends of an aliphatic, 8-atom (9.4A) spacer arm.

Thermo Scientific™ Pierce EMCS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length spacer arm (9.4 Ås).

Highlights:

  • Reactive groups: NHS ester and maleimide
  • Reactive toward: amino and sulfhydryl groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable No
Description EMCS
Molecular Weight (g/mol) 308.29
PEGylated No
Spacer Arm Length 9.4 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, NHS Ester
Spacer Short (<10 Å)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.