missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Fast Western Kit, SuperSignal™ West Pico, Mouse

Product Code. 10605465 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 mL
800 mL
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10605465 200 mL 1 set
10369024 800 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10605465 Supplier Thermo Scientific™ Supplier No. 35065
  • ONLINE EXCLUSIVE PRICE
    190.00 €
    153.90 € / 1 sets
    Save 36.10 €
    19% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For picogram-level detection and Western blot development less than 1 hour after gel transfer.

Thermo Scientific™ Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol that allows picogram levels of detection and shortens typical Western blot development to less than 1 hour after the gel transfer.

These Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Pico Chemiluminescent Substrate.The kits contain all the reagents necessary to complete a Western blot being probed with either a mouse or rabbit primary antibody. Like other Western blotting methods, the included SuperSignal West Pico Substrate produces a chemiluminescent signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.

Highlights:

  • Fast – picogram levels of detection in about one hour
  • Convenient – no expensive hardware or vacuum required; no clogging issues
  • Simple – optimized protocol makes Western blot analysis easier than ever
  • Economical – no additional consumables or extra equipment to purchase
  • Stable – all kit components are stable for one year stored at 4°C
  • Easy – complete kits contain all components needed to block, probe and develop a blot with any standard mouse or rabbit primary antibody

Compatible with:

Pierce Membranes and Filter Paper; CL-Xposure™ Film for Western blotting; Pierce Western Blot Signal Enhancer; Restore Plus™ Western Blot Stripping Buffer

TRUSTED_SUSTAINABILITY

Specifications

Product Type Western Blot Kit
Detection Method Chemiluminescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line Pierce, SuperSignal
Quantity 200 mL
Conjugate HRP
Substrate Horseradish Peroxidase
Reactivity Mouse
Sensitivity Picogram-level
Sufficient For 5 Mini Blots
Content And Storage Sufficient For: 5 mini blots probed with mouse primary antibodies
• Fast Western Antibody Diluent, 100 mL
• Fast Western 10X Wash Buffer, 50 mL
• Fast Western Mouse Optimized HRP Reagent, Pico, 5 mL
• SuperSignal West Pico Luminol/Enhancer Solution, 25 mL
• SuperSignal West Stable Peroxide Solution, 25 mL

Store kit at 4°C.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.