missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FDAA (Marfey's Reagent) (1-fluoro-2-4-dinitrophenyl-5-L-alanine amide)

Product Code. 11561094 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11561094 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11561094 Supplier Thermo Scientific™ Supplier No. TS48895

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Label and measure optical isomers of amino acids by reverse-phase chromatography (HPLC) with this amine-reactive, colored (Amax 340nm) compound.

Thermo Scientific Pierce FDAA (Marfey's Reagents) reacts with primary amines to enable the separation and quantitation of optical isomers of amino acids by reverse-phase chromatography.

Highlights:

  • Optical isomers of amino acids derivatization complete in 90 minutes
  • Derivatives have an absorption coefficient of ∽3 x 104 Derivatives can be detected by UV at 340nm with picomole sensitivity
TRUSTED_SUSTAINABILITY

Specifications

Product Type FDAA
Chemical Reactivity Amine
Content And Storage Upon receipt store product at room temperature.
Label or Dye Other Label or Dye
Quantity 50 mg
Form Solid
compliance-icons
Product Identifier
  • MARFEY'S REAGENT FDAA
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P271-Use only outdoors or in a well-ventilated area.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501-Dispose of contents/container to .

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.