missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Fe-NTA Phosphopeptide Enrichment Kit

Product Code. 10389287 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
30 Column Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10389287 30 Column Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10389287 Supplier Thermo Scientific™ Supplier No. 88300

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purify phosphorylated peptides to process protein digests or cation-exchange fractions for mass spectrometry using spin columns of iron-chelate agarose resin.

Thermo Scientific™ Pierce Fe-NTA Phosphopeptide Enrichment Kit enables fast and efficient enrichment of phosphorylated peptides to process protein digests or strong cation-exchange peptide fractions for analysis by mass spectrometry.

The Pierce Fe-NTA Phosphopeptide Enrichment Kit enriches phosphopeptides from complex samples using iron-chelate resin in spin columns. These columns enrich a higher percentage of phosphopeptides than other resins and with an overall higher number of total and unique phosphopeptides. The complete kit is easy to use and requires less than one hour to process protein digests or strong cation-exchange peptide fractions for analysis by mass spectrometry (MS).

Highlights:

  • Convenient — spin-column format enables parallel processing of multiple samples
  • High capacity — each column enriches up to 150μg of phosphopeptides
  • Excellent recovery — our kit enriches more total and unique phosphopeptides than other commercially available resins
  • High coverage — recover peptides with single and multiple phosphorylation sites

Product Details:

To achieve robust MS results, enrichment of phosphopeptide samples is essential because of low abundance and poor ionization relative to non-phosphorylated peptides. Multiple phosphorylated amino acids within a peptide contribute to the complexity of phosphopeptide analysis. The Pierce Fe-NTA Spin Columns effectively enriches singly phosphorylated peptides as well as peptides with three or more phosphorylated sites, reducing sample complexity and enabling effective identification and characterization of phosphorylated peptides by MS. The Pierce Fe-NTA Phosphopeptide Enrichment Kit contains detailed instructions and all necessary components to load, wash and elute phosphopeptides within an hour. This kit is compatible with samples digested in solution or after in-gel digestion. Using the Pierce Graphite Spin Columns (Part No. 88302) after enrichment will improve phosphopeptide analysis by efficiently binding hydrophilic peptides and efficiently removing urea, salts and other contaminants before MS analysis.

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Fe-NTA Phosphopeptide Enrichment Kit
Formulation Multi-component kit with Fe-NTA Spin Columns
Kit Contents Binding Buffer, 6mL, Elution Buffer, 6mL, Fe-NTA Spin Columns, 30 columns, Wash Buffer (2X), 12mL
Sufficient For Processing 30 samples (200µL each) containing up to 150µg of phosphopeptides each
Quantity 30 Column Kit
For Use With (Application) MALDI-MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.