missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Firefly Luciferase Flash Assay Kit

Product Code. 12192240 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12192240 100 Reaction Kit Each
12102250 1000 Reaction Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12192240 Supplier Thermo Scientific™ Supplier No. 16174
  • ONLINE EXCLUSIVE PRICE
    133.00 €
    107.73 € / Pack of 1
    Save 25.27 €
    19% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Assay regulatory element transcriptional activity in whole cell lysates with this highly sensitive flash-based firefly luciferase kit.

The Thermo Scientific Pierce Firefly Luciferase Flash Assay Kit provides reagents to measure Red firefly luciferase activity in whole cell producing a very strong flash signal which decays rapidly.

Features of the Firefly Luciferase Flash Assay Kit:

  • Highly sensitive detection of firefly luciferase activity
  • Use with Pierce Firefly Luciferase Signal Enhancer (Cat. No. 16180) for best results (maximum signal)
  • Working solution stable for up to 2 months at -20°C
  • Assay reagents compatible with other firefly luciferases
  • Amenable to automation
  • Contains a universal cell lysis buffer and optimized flash assay reagent
  • Safe non-radioactive assay

Thermo Scientific Pierce Firefly Luciferase Flash Assay Kit contains reagents to measure the activity of firefly luciferase in mammalian whole cell lysates. Containing a convenient lysis buffer, assay buffer and firefly substrate, this kit provides a complete reagent system for the intracellular detection of promoter or pathway activity. Light output resulting from the luciferase reaction is captured using a luminometer and can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly expression. These reagents are optimized to work with Thermo Scientific Firefly Luc Plasmids; however, the Flash Assay Kit may be used to detect the activity of other firefly or ATP-dependent luciferases utilizing D-luciferin as a substrate.

Includes:

Cell lysis buffer, reaction buffer and substrate; purchase Luciferase Signal Enhancer (Cat. No. 16180) separately.

Requires:

  • Firefly luciferase and luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers.
  • For best results (maximum signal), use with Pierce Firefly Luciferase Signal Enhancer (Cat. No. 16180)

Applications:

  • Promoter studies for analyzing cis- regulatory elements and trans-acting factors
  • Drug screening
  • siRNA and miRNA screening
  • Multiplexed assays to study off-target effects
  • Secretory pathway/protein localization reporter assays
  • Signal transduction pathway analysis
  • RNA splicing studies

Firefly luciferase is a 60kDa protein produced in nature by several species of the Lampyridae family of beetles which includes the genera Photinus and Luciola. Bioluminescent signal from firefly luciferase originates from the oxidation of D-luciferin. Light output captured using a luminometer can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly expression.

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, Firefly Luciferase
Quantity 100 Reaction Kit
Content And Storage • Firefly Flash Assay Buffer, 5 mL, store at 4°C
• D-Luciferin, Lyophilized, 3 mg, store at 4°C
• 2X Cell Lysis Buffer, 6 mL, store at room temperature

Upon receipt store kit at 4°C or store individual components as indicated above.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer
Format 96-well Plate, 384-well Plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate D-Luciferin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.