missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Formamide

Product Code. 11866714 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11866714 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11866714 Supplier Thermo Scientific™ Supplier No. 17899
Spotlight Offers
Use promo code 28037
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prepare hybridization cocktails and various denaturing solutions used with nucleic acids using this highly pure formulation of formamide denaturant.

Thermo Scientific™ Pierce Formamide is high-purity formulation of formamide denaturant that is suitable for the preparation of hybridization cocktails and various denaturing solutions used with nucleic acids.

Thermo Scientific™  Pierce Formamide is a high-purity formulation of formamide denaturant that is suitable for the preparation of hybridization cocktails and various denaturing solutions used with nucleic acids.

Highlights:

  • Ultra pure formulation possesses a high degree of purity and is free of contaminating RNase and DNase
  • Suitable for the preparation of hybridization cocktails and various denaturing solutions used with nucleic acids
  • Will not cause degradation of expensive or hard-to-isolate RNA and DNA samples
  • Pre-filtered through a mixed-ion exchange resin to eliminate products of decomposition and residual metal cations
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formamide
Content And Storage Store at 2–8°C.
Product Type Formamide
Quantity 100 mL
Form Liquid
compliance-icons
Product Identifier
  • Formamide
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: formamide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.