missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Gaussia-Firefly Luciferase Dual Assay Kit

Product Code. 12201652 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12201652 1000 Reaction Kit Each
12201642 100 Reaction Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12201652 Supplier Thermo Scientific™ Supplier No. 16182
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simultaneously detect intracellular Gaussia and Red Firefly luciferase activity in mammalian whole cell lysates.

The Thermo Scientific Pierce Gaussia-Firefly Luciferase Dual Assay Kit provides the necessary reagents to simultaneously detect intracellular Gaussia and Red Firefly luciferase activity in mammalian whole cell lysates.

Features of the Gaussia-Firefly Luciferase Dual Assay Kit:

  • Simultaneous—concurrent, filter-based, wavelength-separated detection of two luciferase activities
  • Sensitive—measure bright blue Gaussia and red firefly luciferase activities in the same sample
  • Fast—no quenching step required, unlike in traditional sequential dual assays
  • Multiplex—capable of quantitating two cellular activities in the same sample(s)

The Pierce Gaussia-Firefly Luciferase Dual Assay Kit is a highly sensitive assay that allows for the simultaneous detection of Gaussia and Red Firefly luciferase activity. Gaussia luciferase acts as an experimental reporter with constitutively active red firefly as a normalization control. As shown in the animation videos below, this reporter-and-control combination enables simultaneous monitoring of experimental reporter and control luciferase activities in a single-read assay without the need for two-step addition of substrate reagents or quenching. The assay working solution contains substrates for both luciferases, and the reactions occur simultaneously with flash-type kinetics. The resulting luminescent signals are spectrally resolvable using filters. In a single sample, researchers can assay transcriptional activity of regulatory elements, signal transduction pathways, and effects of activators or inhibitors.

Includes:

Cell lysis buffer, reaction buffer, and substrates coelenterazine and D-luciferin

Requires:

Gaussia and Red Firefly luciferase reporters; Filter sets in the ranges of 425 to 525nm for Gaussia Luc and 615 to 675nm for Red Firefly Luc. Luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers; Injector required for assessing more than 24 wells at a time.

Applications:

  • Study two regulatory elements at the same time
  • Monitor two signaling pathways simultaneously
  • Enable studying more than one target per screen (e.g. off-target effects)

Application Videos:

Gaussia luciferase produces brighter bioluminescent signal than firefly and native Renilla luciferases. The bioluminescent signal (λmax= 470nm) produced by Gaussia luciferase results from the oxidation of coelenterazine. This reaction does not require adenosine triphosphate (ATP) or other cofactors. Red firefly luciferase is a mutant form of the Japanese firefly luciferase from Luciola cruciata. This luciferase produces a red-shifted emission spectrum (λmax= 613nm) that results from the oxidation of D-luciferin in the presence of ATP. Red firefly and Gaussia luciferases are used in the dual assay because the light output is spectrally resolvable.

TRUSTED_SUSTAINABILITY

Especificaciones

Type Assay Kit
Target Luciferase, Firefly Luciferase, Gaussia Luciferase
Quantity 1000 Reaction Kit
Content And Storage • Gaussia-Firefly Dual Assay Buffer, 50 mL, store at 4°C
• Coelenterazine (100X), 0.5 mL, store at -80°C
• D-Luciferin, Lyophilized, 30 mg, store at 4°C
• 2X Cell Lysis Buffer, 60 mL, store at room temperature

Upon receipt store kit at -80°C or store individual components as indicated above.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer
Format 96-well Plate, 384-well Plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate Coelenterazine, D-Luciferin
Mostrar más Mostrar menos

For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.