missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Glutathione Agarose

Product Code. 10333058 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
10mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10333058 10 mL 10mL
11884045 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10333058 Supplier Thermo Scientific™ Supplier No. 16100
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purify GST-tagged recombinant fusion proteins from cell lysates using glutathione (GSH) beaded agarose affinity purification resins, spin columns and plates.

Thermo Scientific™ Pierce Glutathione Agarose is a high-capacity, high-performance resin for affinity purification of GST-tagged fusion proteins from cellular lysates.

The high–quality support consists of glutathione that has been immobilized by its central sulfhydryl group via a 12–atom spacer arm to crosslinked 6% beaded agarose. Purification of GST–fusion proteins using glutathione (GSH) agarose beads is well documented and adaptable to a variety of scales, column formats and specific applications. Whether the purpose is to purify large amounts of recombinant protein from over–expressing E. Coli lysates or to investigate protein interactions involving GST–tagged bait proteins, Pierce Glutathione Agarose is suitable for the task.

To accommodate these many uses, Pierce Glutathione Agarose is offered in several package sizes and formats. These include three volumes of resin slurry, three sizes of centrifuge-ready columns, complete GST purification kits, and two sizes of FPLC–ready chromatography cartridges.

Highlights:

  • High capacity – binds at least 25mg of recombinant GST protein per milliliter of resin

  • High yield and purity – consistently purifies at least 10mg of GST – tagged protein per milliliter of resin with greater than 90% purity

  • Cost-effective – resin is economically priced and can be reused at least five times without reduction in binding capacity and purification performance

  • Versatile – works well to purify GST – fusion proteins from bacterial lysates or use with pre-purified GST – tagged proteins to pull down protein interactions

  • Compatible – validated and effective for use with Thermo Scientific Pierce Cell Lysis Reagents to extract and purify from bacterial or mammalian cell cultures

  • Flexible – available in multiple formats including bulk resin, spin columns, chromatography cartridges and convenient GST purification kits

Available Formats:

  • Resin Slurries: 10mL, 100mL, 500mL bottles

  • Spin Columns: 0.2mL (microcentrifuge), 1mL, 3mL columns

  • Purification Kits: complete kits in all three column sizes

  • Chromatography Cartridges: 1mL and 5mL cartridges, compatible with automated liquid chromatography systems and with syringe processing

  • Bulk and Custom: let us know your needs for bulk Glutathione Agarose

Includes:

Kits contain Pierce Glutathione Spin Columns, equilibration/wash buffer and glutathione to prepare elution buffer

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at 4°C.
Product Line Pierce
Quantity 10 mL
Protein Form Recombinant
Type Agarose
Stationary Phase Glutathione
Format Bottle
Column Type Affinity, Agarose Resin
Binding Capacity 40 mg Purified Recombinant GST Protein Binding/mL Resin

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.