Learn More
Description
Thermo Scientific™ Pierce Glycoprotein Staining Kit is a convenient, fast and sensitive kit for staining glycosylated proteins in polyacrylamide gels using the periodic acid-Schiff (PAS) method.
Highlights:
Specific for glycoproteins – periodic acid-Schiff (PAS) method specifically detects glycosylated proteins having sialic acid and other oxidizable carbohydrate groups
Optimized protocol – the traditional three-reagent method has been streamlined to achieve optimal staining in less than two hours (half the time required for other staining methods)
Colorimetric – glycoproteins are detected as magenta bands against a faint pink or colorless gel background
Qualitative staining – detection limit depends on the extent of glycosylation and protein size and abundance; detects 0.625ng avidin and 0.16g horseradish peroxidase per band
Controls included – complete kit includes positive and negative control proteins (HRP and STI, respectively).
Gels and membranes – effective for use with polyacrylamide mini gels or nitrocellulose membranes
Includes:
Three reagents in 250mL bottles (1 solution and 2 dry powders to which water must be added), plus two vials of lyophilized proteins (1mg each)
Specifications
Specifications
| Content And Storage | Sufficient For: 10 mini gels (SDS-PAGE) • Oxidation Reagent, 2.5 g powder to make 250 mL • Reduction Reagent, 1.25 g powder to make 250 mL • Positive Control (Horseradish Peroxidase), 1 mg • Negative Control (Soybean Trypsin Inhibitor), 1 mg • Glycoprotein Stain, 250 mL Store all components at 4°C. |
| Detection Location | In-Blot Detection, In-Gel Detection |
| Detection Method | Colorimetric |
| Label or Dye | Fuchsin Sulfite |
| Quantity | 750 mL |
| Target Molecule | Proteins (Glycoproteins) |
| Product Line | Pierce |
| Product Type | Glycoprotein Staining Kit |
Safety and Handling
- Pierce Glycoprotein Staining Kit
- Danger
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- Carcinogenicity Category 1B
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350-May cause cancer.
- EUH031-Contact with acids liberates toxic gas.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305-IF IN EYES:.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.