missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ GPCR Extraction and Stabilization Reagent

Product Code. 16585651 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16585651 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 16585651 Supplier Thermo Scientific™ Supplier No. A43436
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Enables efficient extraction and stabilization of G-coupled protein receptors (GPCRs) and other membrane-associated proteins from cultured mammalian cells or tissue while preserving longer-term receptor integrity

  • Thermo Scientific GPCR Extraction and Stabilization Reagent enables efficient extraction and stabilization of G-coupled protein receptors (GPCRs) and other membrane-associated proteins from cultured mammalian cells or tissue while preserving longer-term receptor integrity.
  • Ease-of-use—all-in-one formulation to extract and stabilize GPCRs in one hour
  • GPCR preservation—stabilizes the receptor in a detergent micelle to ensure receptor function
  • Optimized—formulation and procedure provide gentle but efficient extraction of GPCRs from the plasma membrane
  • Compatible—lysate is directly compatible with downstream applications including western blotting, immunoprecipitation, and receptor ligand binding assays
  • GPCR Extraction and Stabilization Reagent is simple to use. The sample is first harvested, washed, and pelleted. Receptor is then extracted using the reagent, followed by centrifugation. The resulting whole cell lysate contains solubilized and stabilized receptors. The reagent has been tested for use with endogenous and overexpressed GPCRs, and receptor function has been verified by receptor ligand binding assays.
  • GPCRs are a large family of receptors with seven transmembrane domains that play a key role in transmitting signals across the cell membrane. They are effective drug targets for genetic and immune system disorders and serve as potential biomarkers and therapeutic targets for cancer treatment. One of the major challenges in GPCR research is the ability to efficiently extract the receptor from the membrane without compromising the structural and functional integrity of the protein.
  • GPCR Extraction and Stabilization Reagent encapsulates the receptor in a detergent micelle optimized to ensure receptor function for up to one week when stored at 4°C and one month when stored at -20°C post extraction, providing more flexibility for receptor analysis.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Format Liquid
Product Type Reagent
Volume (Metric) 100 mL
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Room Temperature
Product Line Pierce

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.