missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ActivX™ Desthiobiotin-GTP Probe

Product Code. 11852134 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
16 x 12.9 μg
Unit Size:
Pack of 16
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11852134 16 x 12.9 μg Pack of 16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11852134 Supplier Thermo Scientific™ Supplier No. 88315
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ActivX™ probes have been designed to selectively label and enrich GTPases through binding of their respective active sites with tagged GTP to enable assessment of enzyme activity. Desthiobiotin-GTP can selectively enrich, identify, and profile target enzyme classes in samples.

ActivX™ Desthiobiotin-GTP Probes contain a modified biotin attached to the nucleotide by a labile acyl-phosphate bond. ActivX active-site enrichment probes are advantageous for determining active enzyme levels through the use of analog-tagged probes to allow profiling of both inactive and active enzymes. The ActivX probes selectively enrich only those enzymes that are functionally active and biologically relevant at the time of labeling. With selective enrichment, one can identify and profile target enzyme classes across samples or can assess the specificity and affinity of enzyme inhibitors.

After removal of GTP or GDP nucleotides from enzymes, the desthiobiotin-GTP probe can be used to covalently modify conserved lysine residues in the GTPase nucleotide-binding site. Desthiobiotin-GTP can selectively enrich, identify and profile target enzyme classes in samples. Pre-incubation of samples with small-molecule inhibitors that compete with active-site probes can be used to determine inhibitor binding affinity and target specificity. The ActivX Desthiobiotin-GTP Probe covalently labels the active site of GTPases and GTPase subunits of G-protein coupled receptors.

Applications of ActivX Desthiobiotin-GTP Probe:
• Broad enrichment of GTP-binding proteins from tissues, cells and sub-cellular proteomes
• Enrichment of enzymes based on function
• Profiling of dozens to hundreds of inhibitor targets

Assessment of active-site labeling with desthiobioin probes can be accomplished by either Western blot or mass spectrometry (MS). For the Western blot workflow, desthiobiotin-labeled proteins are enriched for SDS-PAGE analysis and subsequent detection with specific antibodies. For the MS workflow, desthiobiotin-labeled proteins are reduced, alkylated and enzymatically digested to peptides. Only the desthiobiotin-labeled, active-site peptides are enriched for analysis by LC-MS/MS. Both workflows can be used for determining inhibitor target binding, but only the MS workflow can identify global inhibitor targets and off-targets.

These products are subject to a limited use label license.

TRUSTED_SUSTAINABILITY

Specifications

Description ActivX Desthiobiotin-GTP Probe
Content And Storage Store at –80°C
Assay Protein Quantitation Assay
Detection Method Chemiluminescence, Mass Spectrometry, Western Blot
Substrate Type GTPase Substrate
Product Line ActivX
Quantity 16 x 12.9 μg
For Use With (Application) Pull-Down
Target Kinases (Multiple), Phosphodiesterases, ATPases, GTPases, Phosphatases
Type Labeled Nucleotide Probe
Label or Dye Biotin and Other Haptens
Format Tube, Spin Column
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.