missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ HeLa Protein Digest Standard

Product Code. 15235733 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 μg
5 x 20 μg
Unit Size:
20µg
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15235733 20 μg 20µg
15245733 5 x 20 μg Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15235733 Supplier Thermo Scientific™ Supplier No. 88328
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Use this standard, a highly validated mammalian protein digest, as a quality control sample for mass spectrometry (MS) analysis of complex proteomic samples.

Use the Thermo Scientific™ Pierce™ HeLa Protein Digest Standard, a highly validated mammalian protein digest, as a quality control sample for mass spectrometry (MS) analysis of complex proteomic samples.

The Thermo Scientific Pierce HeLa Protein Digest Standard is a highly validated mammalian protein digest that may be used as a quality control sample for mass spectrometry (MS) analysis of complex proteomic samples.

Features of HeLa Protein Digest Standard:

  • Positive control sample—complex mammalian proteome sample protein digest (>15,000 proteins) from the HeLa S3 cell line
  • Thorough tryptic digestion—prepared using both LysC and trypsin to ensure less than 10% tryptic missed cleavages
  • Validated peptide quality—less than 10% methionine oxidation and less than 10% lysine carbamylation
  • Rigorously tested—high-quality, consistent protein digest guaranteed and documented via lot-specific certificates of analysis
  • Stable—provided in a stable lyophilized format

The protein digest standard is derived from a well-established adenocarcinoma reference HeLa S3 cell line. HeLa S3 cells express over 15,000 proteins with relevant post-translational modifications, making this cell line an ideal standard for complex proteome mass spectrometry applications. The cell lysate has been digested with both LysC and trypsin to minimize tryptic missed cleavages and improve protein sequence coverage. Moreover, unlike other commercially available protein digests for MS, the Pierce HeLa Protein Digest Standard meets stringent quality testing specifications, including peptide quality, digestion efficiency, and lot-to-lot digest uniformity.

Applications:

  • Qualitative LC assessment
  • LC/MS standardization
  • LC/MS method development
  • TMT™ Reagent method development

The Pierce HeLa Protein Digest Standard is a lyophilized tryptic peptide mixture that can be used as a quality control standard for liquid chromatography (LC) separation, MS method development and MS performance benchmarking. The digest is specifically formulated for LC/MS experiments and does not contain salts or detergents. Using the digest standard routinely before analysis of complex samples makes it possible to monitor and normalize LC/MS performance between samples and over time.

TRUSTED_SUSTAINABILITY

Specifications

Reagent Type Standard
For Use With (Equipment) Mass Spectrometer
Product Type Protein Digest Standard
Product Line Pierce
Starting Material Peptides
Final Product Type Peptides
Detection Method Mass Spectrometry
Workflow Step MS System Performance
Quantity 20 μg
Form Solid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.