Learn More
Thermo Scientific™ Pierce™ Heptafluorobutyric Acid (HFBA), HPLC Grade
Shop All Thermo Scientific ProductsBeskrivelse
Thermo Scientific™ Pierce Heptafluorobutyric Acid (HFBA) is manufactured and tested to meet strict specifications that ensure superior performance for use as an ion-pairing agent in reverse-phase peptide separations.
Highlights:
- Typical purity is 99.7% by GC; <0.1% water
- Sequencing reagent for classical and automated Edman degradation of peptides and proteins
- Density: 1.645
- B.P. 120°C
- Packaged under nitrogen in amber glass ampules or bottles
- Clear, colorless liquid
Ordering Information:
X in the ordering table indicates that hazardous shipping charges apply.
Tekniske data
Tekniske data
| Item Name | Heptafluorobutyric Acid (HFBA) |
| Content And Storage | Store at room temperature. |
| Form | Liquid |
| Reagent Type | Ion Pairing Reagent |
| Quantity | 10 x 1 mL |
| Formulation | 99.5% HFBA |
| Product Line | Pierce |
For Research Use Only. Not for use in diagnostic procedures.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.