missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High-Capacity EDTA-Compatible Ni-IMAC Resin

Product Code. 17183259 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
2 Plates
250 mL
5 Columns
50 mL
500 mL
Unit Size:
10mL
250mL
500mL
50mL
Pack of 2
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17183259 50 mL 50mL
17173259 10 mL 10mL
17193259 250 mL 250mL
17103269 500 mL 500mL
31759255 5 Columns Pack of 5
31759101 2 Plates Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17183259 Supplier Thermo Scientific™ Supplier No. A50585
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pierce High-Capacity EDTA-Compatible Ni-IMAC Resin is designed for the purification of over-expressed His-tagged proteins from cell culture media.

Pierce High-Capacity EDTA-Compatible Ni-IMAC Resin is designed for the purification of over-expressed His-tagged proteins from cell culture media. The matrix is a 6% crosslinked agarose with optimal properties for batch, column-based, and research/pilot scale FPLC column purifications. The resin has high binding capacity (up to 80 mg of protein per mL of resin) and can tolerate the high EDTA and reducing agent concentrations used to inhibit metalloproteases and stabilize sensitive intracellular proteins.

Features of Pierce high-Capacity EDTA-Compatible Ni-IMAC Resin

  • Chelator stable—use up to 20 mM EDTA and DTT with no loss in performance
  • Versatile—purify both overexpressed and secreted proteins in cell culture media (e.g., Expi293, ExpiCHO, and ExpiSf expression systems)
  • High binding capacity—>80 mg/mL binding capacity for His-tagged recombinant proteins
  • Reusable—loose resin and spin column formats can be regenerated up to 10 times using the standard CIP protocol (not applicable to 96-well plates)
  • Flexible— available in multiple formats including bulk loose resin, spin columns, and 96-well spin plates

EDTA and reducing agents such as DTT and TCEP adversely affect the performance of Ni-IMAC supports by stripping off the metal. Because of its chelator stability characteristic (EDTA tolerant), Pierce High-Capacity EDTA-Compatible Ni-IMAC Resin is specially suited for purification of expressed His-tagged proteins that are secreted into cell culture media or intracellular His-tagged proteins that need the presence of EDTA to maintain stability and function. In both cases, large quantities of samples can be purified without the Ni metal stripping off of the support, thereby providing consistent high-capacity yields.

Available formats

  • Resin slurries—20 mL, 100 mL, 500 mL, and 1 L of 50% slurry in 20% ethanol
  • Spin columns—10 mL spin columns containing 1 mL of settled resin, supplied as 5 mL of 20% slurry
  • 96-well spin plates—96-well spin plates with 50 μL of settled resin, supplied as 250 μL of 20% slurry
TRUSTED_SUSTAINABILITY

Specifications

Binding Property Chelator Stable
Matrix 6% crosslinked agarose
Content And Storage 50 mL settled resin, supplied as 100 mL 50% slurry. Store at 2–8°C.
Shipping Condition Wet ice
Form Liquid Suspension
Product Line Pierce
Quantity 50 mL
Type Resin
Column Type Bottle
Binding Capacity 80 mg/mL Binding
With Pierce High Capacity Ni-IMAC Resin, EDTA compatible, what residence times are adequate for FPLC purification?

Optimized residence times will differ by target, but the standard range is 2-10 mins.

How is Pierce High Capacity Ni-IMAC Resin, EDTA compatible different from standard Ni-NTA resin?

Pierce High Capacity Ni-IMAC Resin, EDTA compatible doesn't leach and discolor in the presence of reducing agent, EDTA, or cell culture supernatant, thereby providing exceptionally high protein yield.

After regeneration of Pierce High Capacity Ni-IMAC Resin, EDTA compatible, is there a specific buffer the resin should be stored in?

The recommended storage buffer for the column and resin is 20% ethanol, 10 mM sodium acetate, pH 6.50. Please refer to the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0024862_PierceHighCapNiIMACResin_UG.pdf) for details.

With Pierce High Capacity Ni-IMAC Resin, EDTA compatible, how do I regenerate the resin?

Pierce High Capacity Ni-IMAC Resin, EDTA compatible is alkali-stable. The resin can be exposed to NaOH, washed with water, neutralized with PBS, washed with water, and put back into storage buffer. Please refer to the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0024862_PierceHighCapNiIMACResin_UG.pdf) for details.

How many uses/regenerations are possible with Pierce High Capacity Ni-IMAC Resin, EDTA compatible?

Up to 10 uses are possible with Pierce High Capacity Ni-IMAC Resin, EDTA compatible.

What flow rates can I use with Pierce High Capacity Ni-IMAC Resin, EDTA compatible?

With Pierce High Capacity Ni-IMAC Resin, EDTA compatible, we recommend using a flow rate of 0.5-1.0 mL/min, but can go up to 2 mL/min depending on the sample type.

What is the binding capacity of Pierce High Capacity Ni-IMAC Resin, EDTA compatible?

The binding capacity of Pierce High Capacity Ni-IMAC Resin, EDTA compatible is up to 60 mg/mL.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.