missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hydrochloric Acid Solution, Sequencing Grade

Product Code. 11541074 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11541074 10 x 1 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11541074 Supplier Thermo Scientific™ Supplier No. TS24308
Quantity Price
1 63.44 € / Pack of 10
10 56.22 € / Pack of 10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Constant-boiling 6N HCl solution, supplied in 1mL ampules, used to acid-hydrolyze proteins (protein hydrolysis) for HPLC amino acid composition analysis.

Thermo Scientific™ Pierce Hydrochloric Acid Solution is a constant-boiling 6N HCl solution, supplied in 1mL ampules, that is specifically formulated as a sequencing-grade reagent for total protein hydrolysis for amino acid analysis.

Highlights:

  • Hydrolyzes peptides in 6 hours at 150°C
  • Specially purified to give ninhydrin-negative blank on hydrolysis
  • Packaged in prescored ampules to eliminate contamination and ensure product integrity

Ordering Information:

X in the ordering table indicates that hazardous shipping charges apply.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Hydrochloric Acid Solution
Content And Storage No product PDF available for storage information.
Quantity 10 x 1 mL
Form Solid
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric Acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.