missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Immunodiffusion Plates, Multiple Pattern

Product Code. 10036373 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
6 plates
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10036373 6 plates Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10036373 Supplier Thermo Scientific™ Supplier No. 31111
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Identify and characterize antibody-antigen binding interactions with these standard Ouchterlony gel plates with patterns of six wells around one well.

Thermo Scientific™ Pierce Immunodiffusion Plates are standard Ouchterlony gel plates containing patterns of six wells around one well for studying and characterizing antibody-antigen binding interactions.

Immunodiffusion (ID) is a classic technique for detecting antibody-antigen interactions based on the theory of double diffusion originally described by Oudin1 and Ouchterlony.2 Antigens and antibodies are placed into separate wells that are cut into a gel matrix and allowed to diffuse towards each other. If the reaction is positive, a precipitate forms that appears as an opaque line. The precipitation reaction occurs when the antigen and antibody concentrations are combined at near equivalent proportions.

When multivalent antigens combine with divalent antibodies in solution, three-dimensional lattices are formed that aggregate and precipitate. The amount of precipitate varies in proportion to the concentration of the antigen and antibody. At equivalent or optimal proportions almost all the antigen and antibody will precipitate. If there is an excess of antibody, the complexes formed with the antigen are insoluble. When there is an excess of antigen, the precipitate has a tendency to dissolve caused by the formation of soluble complexes.

Highlights:

  • Gelling agent contains precipitin brighteners and diffusion enhancers that help visualize and speed the interaction process, respectively
  • Good precipitin bands from all species (including rabbit)
  • Gels can be washed, dried and stained for a permanent record
TRUSTED_SUSTAINABILITY

Specifications

Assay Immunodiffusion (ID)
Content And Storage Upon receipt store at 4°C. Plates are shipped at ambient temperature. Do not freeze plates or allow them to become dry.
Detection Method Precipitate
Color Clear
Type Multi-Well Plate
Volume (Metric) Working 22 μL
Well Count 6
Product Line Pierce
Quantity 6 plates
Target Specificity Not Target-Specific
Technique Immunodiffusion (ID)
Height (Metric) 2.8 mm
Diameter (Metric) 3.2 mm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.