missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Intact Protein Standard Mix

Product Code. 15786003 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 76 μg
5 x 76 μg
Unit Size:
76µg
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15786003 5 x 76 μg Pack of 5
15776003 1 x 76 μg 76µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 15786003 Supplier Thermo Scientific™ Supplier No. A33527
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Intact Protein Standard Mix
Product Line Pierce
Content And Storage 5 vials, store at -5 to -30°C
Shipping Condition Wet Ice
Workflow Step Calibration
Quantity 5 x 76 μg
Form Powder

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.