missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Lane Marker Non-Reducing Sample Buffer

Product Code. 11819340 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11819340 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11819340 Supplier Thermo Scientific™ Supplier No. 39001
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Ready-to-use 5X SDS-PAGE sample loading buffers available in reducing and nonreducing formulations, both with a pink dye buffer-front marker.

Thermo Scientific™ Pierce Lane Marker Sample Buffers are ready-to-use 5X SDS-PAGE sample loading buffers, available in reducing and nonreducing formulations, with a pink dye buffer-front marker.

Lane Marker Reducing and Non-Reducing Sample Buffers are convenient and ready to use for SDS-polyacrylamide gel electrophoresis (SDS-PAGE) and Western blotting. Formulated as 5X stocks rather than the traditional 2X stocks, they enable a larger volume of protein solution to be included in the sample that is loaded in each well. Particularly unique in these buffers is the use of a bright pink tracking dye instead of the traditional bromophenol blue dye. This pink dye marks not only the dye front in the gel but also transfers and permanently binds to nitrocellulose membranes. Consequently, the dye can be used to assess transfer efficiency and align cut membranes. The Reducing Sample Buffer contains DTT as the reducing agent, eliminating the repugnant odor associated with mercaptoethanol-containing buffers.

Lane Marker Sample Buffers are easy to use. Simply mix one part Sample Buffer with four parts protein sample, heat denature, and then load the gel sample wells. Visualize progress of the electrophoresis run by monitoring the location of the pink dye front. Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with coomassie dye and silver staining techniques, as well as Western blotting procedures.

Highlights:

  • Easy to see – bright pink dye marks the buffer front, clearly indicating the progress of the electrophoresis run

  • Concentrated – 5X stocks enable a greater range of protein sample volumes to be used for electrophoresis

  • Transferable marker dye – pink dye transfers with protein to nitrocellulose membrane, enabling verification of transfer direction and orientation of the blot

  • Flexibility in SDS-PAGE applications – available in two ready-to-use formulations for reducing or non-reducing denaturing SDS polyacrylamide electrophoresis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at room temperature. Product is shipped at ambient temperature.
Description Pierce Lane Marker Non-Reducing Sample Buffer (5X)
Buffer Sample Loading Buffers
Concentration 5X
Detection Method Fluorescence
Gel Type SDS-PAGE
Gel Compatibility SDS-PAGE Gels
Quantity 5 mL
Product Line Pierce
Product Type Lane Marker Non-Reducing Sample Buffer
compliance-icons
Product Identifier
  • Lane Marker Non-Reducing Sample Buffer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: glycerol, sodium dodecyl sulphate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.