missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Magnetic RNA-Protein Pull-Down Kit

Product Code. 13444209 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 reactions kit
Unit Size:
20 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13444209 20 reactions kit 20 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13444209 Supplier Thermo Scientific™ Supplier No. 20164
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Utilize a streamlined, robust method to enrich protein-RNA interactions using end-labeled RNA as the bait with Thermo Scientific™ Pierce Magnetic RNA-Protein Pull-Down Kit.

Utilize a streamlined, robust method to enrich for protein-RNA interactions using end-labeled RNA as the bait with Thermo Scientific™ Pierce Magnetic RNA-Protein Pull-Down Kit.

Additional Information and Online Ordering

The Magnetic RNA-Protein Pull-Down Kit provides reagents to efficiently enrich RNA Binding Proteins (RBPs) using RNA end-labeled with desthiobiotin and streptavidin magnetic beads. The complete kit contains sufficient reagents for 20 RNA labeling reactions and 20 protein-RNA pull-down assays. This direct enrichment of the protein-RNA interaction provides an alternative to antibody capture of protein-RNA complexes or moieties incorporated into the nucleic acid. An added advantage to the kit is that it includes validated controls for both the labeling and pull-down assay. The kit is amenable to several downstream applications, including Western blotting and Mass Spectrometry (MS).

Highlights:

  • Direct – capture ribonucleoprotein complexes directly with end-labeled RNA; does not use or require antibodies for pull-down
  • Easy to use – no spin cups or centrifugation necessary for the enrichment of RBP; procedure streamlined for minimal hands-on time (less than 3 hours) after RNA labeling reaction
  • Flexible – use in vitro transcribed RNA or synthetic RNA for labeling of various lengths and complexity; proteins successfully enriched using endogenous, over-expressed, and in vitro translated lysates
  • Specific – low bead background; unrelated RNA or mutated RNA does not significantly enrich specified RBPs
  • Economical – less expensive than purchasing commercially synthesized end-labeled RNA, magnetic beads, and reagents separately
  • Complete – contains both labeling and enrichment modules with buffers necessary for assay; positive control RNA, negative control RNA, and RBP antibody included

Includes:

Complete kit contains the Pierce RNA 3'-End Desthiobiotinylation Kit, positive and negative RNA controls, nucleic-acid compatible streptavidin magnetic beads, and buffers for RBP enrichment and elution

Requires:

User-supplied RNA and lysate (experimental sample)

Recommended for:

  • Mutational analysis
  • Structure function analysis
  • Identification of RNA:Protein interactions
  • MS analysis of unknown RNA:protein binding pairs or complexes
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 mL
Kit Contents 50 pmol of RNA per Reaction
Content And Storage Sufficient For: 20 RNA-protein complex pull-down reactions

Slurry: 10mg/mL, 1% solids
• Pierce Nucleic Acid-Compatible Streptavidin Magnetic Beads, 1 mL (store at 4°C)
• RNA Capture Buffer (1X), 10 mL (store at 4°C)
• Tris (20 mM, pH 7.5), 5 mL (store at 4°C)
• Protein-RNA Binding Buffer (10X), 1 mL (store at 4°C)
• Wash Buffer (1X), 10 mL (store at 4°C)
• Biotin Elution Buffer, 1.5 mL (store at 4°C)
• HuR Monoclonal Antibody (Mouse), 50 μL (store at 4°C)
• Positive RNA Control (AR RNA), 250 pmol (store at -20°C)
• Negative RNA Control (polyA25 RNA), 250 pmol (store at -20°C)
• Pierce RNA 3' End Desthiobiotinylation Kit (part number 20163; store at -20°C)
Product Line Pierce
Quantity 20 reactions kit
Type RNA-Protein Pull Down Kit
Sufficient For 20 Reactions
Format Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.