missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 16% Formaldehyde (w/v), Methanol-free

Product Code. 11586711 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 1 mL
10 x 10 mL
Unit Size:
100mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11586711 10 x 10 mL 100mL
10751395 10 x 1 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11586711 Supplier Thermo Scientific™ Supplier No. 28908

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vials of high-quality, methanol-free formaldehyde, a general IHC fixative and reversible amine-crosslinking agent for protein and nucleic acid samples.

Thermo Scientific™ Pierce Formaldehyde Ampules are vials of high-quality, methanol-free formaldehyde for use as a general immunohistochemistry fixative and reversible amine-crosslinking agent for protein and nucleic acid tissue samples.

Formaldehyde is a highly reactive, cell-permeable agent that is used as a reversible cross-linking agent for proteins and nucleic acids within the cell or as a general cell-fixing agent for imaging-based applications. Cell permeability allows the reagent to readily enter living cells, enabling intracellular applications, such as protein interaction discovery or bioimaging. Formaldehyde produces reversible cross-links with protein and nucleic acids by coupling neighboring primary amines. Cross-links can be reversed simply by heating the sample.

Highlights:

  • Pure and stable - prepared directly from pure granular paraformaldehyde and sealed in ampules under an inert atmosphere, ensuring that it is methanol-free and resistant to oxidation

  • Versatile - use to cross-link molecules in protein-protein and protein-nucleic acid interaction discovery, bioimaging and flow cytometry

  • Aliquot packaging - choose 1mL or 10mL ampules suitable for low- and high-volume applications ranging from culture-targeted cell fixing to in-cell protein cross-linking

  • Convenient starting concentration - 16% (w/v) formaldehyde solution in ultrapure water simplifies dilution to the required working concentration

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formaldehyde
Quantity 10 x 10 mL
Physical Form Liquid
Product Line Pierce
compliance-icons compliance-icons
Product Identifier
  • 16% Formaldehyde Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P201-Obtain special instructions before use.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.