missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MA(PEG)8 Methyl-PEG-Amine Compound

MA(PEG)n: methyl- and amine-capped polyethylene glycol compounds containing 4 to 24 PEG units; use for surface-modification and molecule-pegylation applications.

Brand:  Thermo Scientific™ 26113

Code : A0

Additional Details : Weight : 0.00100kg Transport : UN number : 3261 Chem class : 8 Pack group : III

 View more versions of this product

Product Code. 10045893

  • 1178.00 € / 1g

Estimated Shipment: 07-04-2023
to see stock.
Add to basket

Description

Description

Thermo Scientific™ Pierce MA(PEG)n Compounds are methyl- and amine-terminated polyethylene glycol reagents, where n equals 4 to 24 PEG units, that are useful for a variety of surface-modification and molecule-pegylation applications.

MA(PEG)n compounds have the form Methyl-PEGn-Amine and are ideal for derivatizing surfaces and molecules with discrete numbers (n = 4, 8, 12, 24) of ethylene glycol groups. Use any amine-reactive chemistry (e.g., EDC or NHS ester) to PEGylate surfaces, proteins, or other molecules with a precise number of hydrophilic, nonimmunogenic PEG units.

Highlights:

  • Pure compounds of specific size ensure complete control of PEGylation
  • Straight-chain compounds with simple terminal primary amine
  • Adaptable to a variety of modification procedures

Recommended for:

  • PEGylate carboxylate or amine surfaces Add inert mass to proteins, immunogens, drug compounds and probes Improve solubility (decrease aggregation) of proteins or peptides without affecting function Protect proteins from proteolysis
Specifications

Specifications

Carboxylic Acid, Ketone, Aldehyde
Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature.
Amine, Primary Amine
Solid
Water
Pierce MA(PEG)8
Polyethylene Glycol (PEG), Blocking Groups
8 ethylene glycol units
1 g
Amine Compound
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers