missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Micro BCA™ Reagent C (MC)

Product Code. 10753505 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
12 mL
Unit Size:
12mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10753505 12 mL 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10753505 Supplier Thermo Scientific™ Supplier No. 23234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Special 3-component version of our popular BCA Reagents to measure total protein concentration of dilute protein solutions (0.5 to 20 micrograms/mL).

Thermo Scientific™ Pierce Micro BCA Protein Assay uses a concentrated form of the BCA Reagents to accurately measure dilute protein solutions (0.5 to 20μg/mL).

Additional Information and Online Ordering

The Micro BCA Protein Assay Kit is a specialized version of the popular Pierce BCA Protein Assay for determining the protein concentration of dilute samples. Mixing together the three Micro BCA Reagents results in a working solution that is sufficiently concentrated to measure protein when mixed with an equal volume of sample. The result is an assay for accurately measuring 0.5 to 20μg/mL protein solutions. The assay is exceptionally linear and exhibits very low levels of protein-to-protein variability.

Highlights:

  • Sensitive – accurately detect down to 0.5μg/mL (2μg/mL in microplate format)

  • High linearity – linear working range for BSA equals 0.5 to 20μg/mL

  • Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)

  • Compatible – unaffected by typical concentrations of most ionic and nonionic detergents

  • Convenient – microplate and cuvette protocols provided with the instructions

  • Stable – kit stable at room temperature; prepared working reagent stable for to 24 hours

Includes:

Three Micro BCA Reagents (MA, MB, MC) and 10 ampules of Albumin Standard (2mg/mL)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store product at room temperature. Product shipped at ambient temperature.
Description Pierce Micro BCA Reagent C (MC)
Assay BCA Assay
Detection Method Colorimetric
Product Line Micro BCA, Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
Sufficient For 3200 Microplate Assays
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 12 mL
Show More Show Less
compliance-icons
Product Identifier
  • Micro BCA Reagent C (MC)
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : Copper (II) sulfatepentahydrate (1:1:5)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.