missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ MS-Compatible Magnetic IP Kits

Product Code. 15312627 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Protein A/G Magnetic Beads
Streptavidin Magnetic Beads
Unit Size:
40 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
15312627 Protein A/G Magnetic Beads 40 reactions
15302627 Streptavidin Magnetic Beads 40 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 15312627 Supplier Thermo Scientific™ Supplier No. 90409

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Achieve highly efficient, compatible immunoprecipitation and co-immunoprecipitation of target antigens for downstream LC-MS analysis using Thermo Scientific Pierce MS-Compatible Magnetic IP Kits with streptavidin or Protein A/G magnetic beads.

Immunoprecipitation is often necessary for the detection and quantitation of low-abundance proteins or rare signaling events. Mass spectrometry analysis relies on two methods for the detection of low-abundance proteins: single reaction monitoring (SRM) and parallel reaction monitoring (PRM), which enable isolation, scheduling, and quantitation of selected proteotypic peptides. These IP kits have been optimized for efficient immunoprecipitation and co-immunoprecipitation of target proteins, and are directly compatible with downstream MS sample preparation and analysis.

MS-compatible—reagents directly compatible with in-solution peptide digestion
Sensitive—procedure successfully enriches low-abundance proteins (low ng range)
Low background—enriched samples contain minimal detergent residuals
Reduced antibody contamination—antibody biotinylation with streptavidin enrichment reduces contamination in the eluate when compared to other IP methods
Robust—procedure and reagents have been verified with numerous targets to ensure consistent enrichment of low-abundance proteins (ng range) with at least two peptides identified per protein

The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Protein A/G) uses high-quality Pierce Protein A/G Magnetic Beads to provide wider flexibility of antibody capture than either Protein A or G alone. The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Streptavidin) uses high-quality Pierce Streptavidin Magnetic Beads for use with biotinylated antibodies that may be purchased commercially or labeled using the Pierce Antibody Biotinylation Kit for IP. After the immunoprecipitation procedure, the target-enriched elution fraction is ready for in-solution tryptic digestion for LC-MS analysis without the need for gel purification, detergent removal, or desalting. Additionally, the reagents and procedures have been validated using both manual and automated magnetic separation.

Applications
• Enrichment of low-abundance targets to improve MS sensitivity
• Identification of target proteins that are not detectable by western blotting
• Quantitation of target proteins by LC-MS analysis
• Screening of antibodies for IP-MS

TRUSTED_SUSTAINABILITY

Specifications

Description Protein A/G Magnetic Beads
For Use With (Equipment) KingFisher™ Flex, Magnetic Stand
Product Type Magnetic IP Kit
Product Line Pierce
Sufficient For 40 Reactions
Content And Storage Protein A/G Magnetic Beads, 1 mL
IP-MS Cell Lysis Buffer, 100 mL
IP-MS Wash Buffer A, 75 mL
IP-MS Wash Buffer B, 40 mL
IP-MS Elution Buffer, 6 mL
MgCl2 (1 M), 0.75 mL

Store in refrigerator (2° to 8°C).
Starting Material Protein Samples, Cell Lysate
Final Product Type Protein
Detection Method Mass Spectrometry
Format Kit
Workflow Step Sample Enrichment
Quantity 40 reactions kit
For Use With (Application) Mass Spectrometry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.