missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NBT Substrate Powder (nitro-blue tetrazolium chloride)

Product Code. 10046213 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10046213 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10046213 Supplier Thermo Scientific™ Supplier No. 34035
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Powders and ready-made nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate solutions for chromogenic detection of alkaline phosphatase probes.

Thermo Scientific™ Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.

The combination of NBT (nitro-blue tetrazolium chloride) and BCIP (5-bromo-4-chloro-3'-indolyphosphate p-toluidine salt) yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, a popular enzyme conjugate for antibody probes. Pierce NBT and BCIP powders are available individually and in two convenient, ready-to-use solutions (with and without a levamisole suppressor of endogenous phosphatase activity).

Highlights:

  • AP substrate – for detection of alkaline phosphatase (alk-phos) activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples

  • Chromogenic – no special equipment needed for visualization; produces purple-black precipitate that is easy to photograph

  • Package options – choose individual NBT and BCIP powders, or preformulated, ready-to-use solutions with or without levamisole suppressor

1-Step™ NBT/BCIP

  • Ideal for immunoblotting applications
  • Minimal assay-to-assay variability

1-Step NBT/BCIP Plus Suppressor

  • Ideal for staining tissues assayed with calf intestinal alkaline phosphatase without the interference caused by endogenous phosphatases
  • Contains 1mM levamisole to inhibit endogenous phosphatase activity
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce NBT Substrate
Product Type Substrate
Detection Method Colorimetric
Form Powder
Membrane Compatibility Nitrocellulose, PVDF
Quantity 1 g
Substrate Alkaline Phosphatase
Reactivity Alkaline Phosphatase
Content And Storage Upon receipt store product at 4°C protected from moisture. Product is shipped at ambient temperature.
What alkaline phosphatase substrates do you offer?
compliance-icons
Product Identifier
  • NBT Substrate Powder (nitro-blue tetrazolium chloride)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.