Learn More
Thermo Scientific™ Pierce™ NHS-Activated Agarose, Dry
Shop All Thermo Scientific ProductsDescription
Perform rapid and stable immobilization of proteins, peptides and other ligands with Thermo Scientific™ Pierce NHS-Activated Agarose, a high-quality beaded-agarose support.
The Pierce NHS-Activated Agarose resin is available in an anhydrous acetone slurry and as dry powder. The unique dry form does not require storage in or removal and disposal of the acetone solvent. In addition, the dry resin is ideal for coupling reactions with dilute samples because it concentrates the sample as the resin swells, reducing the volume of the starting material and resulting in highly efficient ligand immobilization.
Highlights:
Easy to use – immobilize in a simple one-step reaction with minimal hands-on time
Rapid and efficient – greater than 85% coupling for most proteins within 30 minutes
Innovative – the dry agarose concentrates the sample, making it ideal for immobilizing dilute proteins
Safe – no hazardous chemicals needed (e.g., sodium cyanoborohydride, cyanogen bromide)
Versatile – affinity resin is adaptable to column and batch affinity chromatography techniques and FPLC applications
Compatible – use with any primary amine-containing compound
Reusable – the leak-resistant chemistry means you can reuse the affinity resin
High binding capacity – binding capacity of >30mg/mL (slurry) or >25mg/mL (dry resin)
Specifications
Specifications
| Capacity (Metric) | ≥25 mg per mL Coupling |
| Content And Storage | Upon receipt store dry resin at -20°C. Product is shipped in an ice pack. |
| Product Line | Pierce |
| Quantity | 1 g |
| Protein Form | Recombinant |
| Target | Antibody, Protein, Peptide, Ligand |
| Type | Agarose |
| Stationary Phase | NHS Ester |
| Format | Vial |
| Column Type | Affinity, Agarose Resin |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.