missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SDAD (NHS-SS-Diazirine) (succinimidyl 2-((4,4'-azipentanamido)ethyl)-1,3'-dithiopropionate)

Product Code. 11811425 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11811425 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11811425 Supplier Thermo Scientific™ Supplier No. 26169
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Very short to mid-length (3.9 to 13.5A), amine-to-anything crosslinkers; NHS-ester and diazirine reactive groups; photoactivate with UV light (Amax 330 to 370nm).

Thermo Scientific™ Pierce NHS-Diazirine Crosslinkers combine proven NHS-ester and diazirine-based photoreaction chemistries to conjugate amine-containing molecules with nearly any other functional group via long-wave UV-light activation.

The succinimidyl-ester diazirine (SDA) reagents are a new class of crosslinkers that combine proven amine-reactive chemistry with an innovative and efficient diazirine-based photochemistry for conjugating amine-containing molecules to nearly any other functional group. The SDA crosslinkers include six compounds differing in spacer arm lengths, ability to cleave the crosslinked proteins, and presence or absence of a charged group for membrane permeability. Protein crosslinking is an important technique used to understand protein structure and to stabilize protein-protein interactions. Thermo Scientific SDA Reagents extend the efficiency and range of interactions that can be explored by this approach.

Highlights:

  • Versatile - different amine-reactive diazirine derivatives capture specific intracellular or cell-surface protein interactions

  • Controllable - two-step chemical crosslinking is activated using common laboratory UV lamps

  • Reversible - disulfide-containing spacers enable cleavage using reducing agents

  • Easy - reagents are photo-stable under typical laboratory lighting conditions so there is no need to perform experiments in the dark

  • Better than aryl azides - diazirine photo-reactive group has better photostability and is more efficiently activated by long-wave UV light

TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Nonselective
Cleavable By Thiols
Description SDAD
Molecular Weight (g/mol) 388.46
PEGylated No
Spacer Arm Length 13.5 Å
Content And Storage Upon receipt store product at 4°C protected from light and moisture.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety NHS Ester, Diazirine
Spacer Medium (10 to 30 Å)
Form Solid
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.