missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ OPD (o-phenylenediamine dihydrochloride)

Product Code. 11869250 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11869250 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11869250 Supplier Thermo Scientific™ Supplier No. 34005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Water-soluble horseradish peroxidase (HRP) substrate that produces a yellow-orange product detectable at 492nm by ELISA plate readers.

Thermo Scientific™ Pierce OPD is a water-soluble substrate for horseradish peroxidase (HRP) that produces a yellow-orange product detectable at 492nm by ELISA plate readers.

OPD (o-phenylenediamine dihydrochloride) yields a water-soluble yellow-orange product when reacted with peroxidase with an absorbance maximum of 492nm. OPD can easily be dissolved in a substrate buffer such as Thermo Scientific Stable Peroxide Substrate Buffer (Part No. 34062).

Highlights:

  • Water soluble product when reacted with peroxidase
  • Absorbance maximum of 492nm
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce OPD Powder
Product Type ELISA Substrate
Quantity 25 g
Detection Method Colorimetric
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)
Content And Storage Upon receipt store product at -20°C. Product is shipped at ambient temperature.
compliance-icons compliance-icons
Product Identifier
  • OPD (o-phenylenediamine dihydrochloride)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: o-phenylenediamine dihydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.