missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lysosome Enrichment Kit for Tissues and Cultured Cells

Product Code. 10696574 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
230 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10696574 230 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10696574 Supplier Thermo Scientific™ Supplier No. 89839

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Enrich for lysosomes, peroxisomes and nuclei from cells and tissue with these organelle-specific subcellular fractionation kits.

Perform enrichment of intact organelles including lysosomes, peroxisomes and nuclei from cells and tissue with Thermo Scientific™ Organelle Enrichment Kits.

These three organelle-specific kits provide the media and reagents necessary to accomplish subcellular fractionation using high-speed density gradient centrifugation. Subcellular fractionation simplifies otherwise complex protein mixtures that result from whole cell lysis, thereby facilitating proteomic analysis of specific organelle targets and pathways. Each kit uses density gradient centrifugation to separate organelles from contaminating cellular structures. The isolated organelles may be used for a number of downstream applications, including 2D/MS, electron microscopy, disease profiling, gene expression, signal transduction and interaction or localization studies.

Highlights:

  • Efficient and easy to use – kit reagents and gradient centrifugation separate organelles from contaminating structures

  • Compatible – prepare samples for downstream applications, including 2D/MS, electron microscopy, disease profiling, gene expression, signal transduction and interaction or localization studies

  • Complete – kits contain sufficient material for 25 applications

Includes:

Organelle-specific Reagents A and B, Optiprep™ Cell Separation Media, and phosphate-buffered saline

Requires:

Ultracentrifuge, refrigerated microcentrifuge, vortex mixer, Dounce tissue grinder, sonicator

TRUSTED_SUSTAINABILITY

Specifications

Product Type Lysosome Enrichment Kit
Content And Storage Upon receipt store at 4°C. Product shipped at ambient temperature
Form Liquid
Reagent Type Organelle Isolation, Subcellular Fractionation
Sufficient For 25 Samples of 50 to 200 mg Cells or Tissues Each
Quantity 230 mL
compliance-icons
Product Identifier
  • Lysosome Enrichment Kit for Tissues and Cultured Cells
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium chloride, Sodium phosphate dibasic

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.