missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PEG4-SPDP (PEGylated, long-chain SPDP crosslinker)

Product Code. 11802164 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11802164 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11802164 Supplier Thermo Scientific™ Supplier No. 26128
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Long (25.7A) amine-to-sulfhydryl crosslinker; NHS-ester and pyridyldithiol groups at ends of 4-unit polyethylene glycol spacer arm; forms cleavable disulfide bonds; water-soluble.

Thermo Scientific™ Pierce PEG4-SPDP is a multifunctional crosslinker for protein conjugation via amine-to-amine or amine-to-sulfhydryl crosslinks that contain a 4-unit polyethylene glycol (PEG) group and a reducible (cleavable) disulfide bond.

SPDP reagents have an amine-reactive N-hydroxysuccinimide (NHS) ester at one end and a suflhydryl-reactive 2-pyridyldithio group at the opposite end. PEG4-SPDP has a 4-unit polyethylene glycol spacer arm, which confers greater solubility to the crosslinker and linked proteins compared to crosslinkers having only hydrocarbon spacers. Pyridyldithiol reagents produce disulfide-containing linkages that can be cleaved with reducing agents such as dithiothreitol (DTT). Crosslinking experiments with SPDP reagents are not limited to those involving proteins. Any of a variety of molecules with primary amines and sulfhydryl groups can be modified or crosslinked using an SPDP reagent.

Highlights:

  • Efficient amine conjugation – NHS ester provides effective conjugation to lysines, N-termini of peptides and other primary amines

  • Reversible sulfhydryl conjugation – pyridyldithio group enables disulfide linkage with reduced cysteines and other sulfhydryl groups, providing a stable but cleavable bond

  • Polyethylene glycol – PEG groups are flexible, non-immunogenic, hydrophilic, and often enhance the solubility of attached molecules

  • Spacer arm – connects primary amines and sulfhydryl targets at distances up to 25.7 angstroms

Specifications:

  • Chemical name (simplified): 2-pyridyldithiol-tetraoxatetradecane-N-hydroxysuccinimide
  • Chemical formula: C23H33N3O9S2
  • Molecular weight: 559.65
  • Spacer arm: 25.7 angstroms
  • Form: straw to amber colored oil
  • Solubility: dissolve 2mg in 179μL DMSO or DMF to make 20mM stock for each use
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Description PEG4-SPDP
Molecular Weight (g/mol) 559.65
PEGylated Yes
Spacer Arm Length 25.7 Å
Content And Storage Upon receipt store at -20°C protected from moisture.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety NHS Ester, Pyridyldisulfide
Spacer Medium (10 to 30 Å)
Form Liquid
Quantity 100 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.