missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Peptide Retention Time Calibration Mixture

Product Code. 11310360 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11310360 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11310360 Supplier Thermo Scientific™ Supplier No. 88321
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

15 synthetic heavy peptides mixed in equimolar ratio to test and calibrate reverse-phase liquid chromatography (LC) and mass spectrometry (LC-MS) equipment.

Thermo Scientific™ Pierce Peptide Retention Time Calibration Mixture enables reversed phase liquid chromatography (LC) and mass spectrometry (LC-MS) users to optimize and confirm correct operation of their equipment.

The Pierce Peptide Retention Time Calibration Mixture is used to optimize and assess LC parameters, identify total peptide elution window and optimize MS parameters. Testing the mixture frequently throughout the lifetime of a column or including it in quantitative samples makes it possible to monitor and normalize LC-MS performance between samples and over time. When used with Thermo Scientific Pinpoint 1.1 software, the calibration mixture is used to predict peptide retention times based upon peptide sequence and ease the transfer of methods between LC-MS platforms.

Highlights:

  • Assessment of chromatography and MS instrument performance
  • Prediction of peptide retention across multiple instrument platforms
  • Prediction of peptide retention time from sequence using calculated hydrophobicity factor
  • Optimization of scheduled MS acquisition windows for improved quantification and increased multiplexing
  • Internal standard to normalize for variation in retention times and peak intensities between runs
TRUSTED_SUSTAINABILITY

Specifications

Formulation Mixture of 15 synthetic heavy peptides at 5uM each
For Use With (Equipment) Mass Spectrometer
Product Type Peptide Retention Time Calibration Mixture
Product Line Pierce
Content And Storage Upon receipt store at -80°C.
Starting Material Peptides
Final Product Type Peptides
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 200 μL
Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Pierce Peptide Retention Time Calibration Mixture
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: acetonitrile

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.