missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Peroxidase Suppressor

Product Code. 10177493 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10177493 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10177493 Supplier Thermo Scientific™ Supplier No. 35000
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Stable, easy-to-use endogenous peroxidase inhibitor to reduce background signal from cells and tissues in HRP-based IHC staining.

Thermo Scientific™ Pierce Peroxidase Suppressor is a stable, easy-to-use inhibitor of endogenous peroxidase activity from cells and tissues to reduce background signal in HRP-based immunohistochemistry applications.

The immunoperoxidase method is a commonly used detection method for immunostaining of cells and tissues. However, the analysis often is complicated by the presence of endogenous peroxidase and “pseudoperoxidase” activity in the cells and tissues. This presence results in nonspecific staining when a substrate such as diaminobenzidine (DAB) is present. Nonspecific staining is difficult to distinguish from the immunochemical marker, making it difficult to distinguish the final location of the antigen of interest. The typical method of inhibiting endogenous peroxidase activity involves hydrogen peroxide or hydrogen peroxide in methanol. The peroxidase suppressor inhibits peroxidase activity even more effectively than the method using hydrogen peroxide in methanol. Plus, the Peroxidase Suppressor does not affect antibody activity. Peroxidase Suppressor also can be used on blots of whole cell supernatant prior to the addition of the horseradish peroxidase (HRP) marker.

Highlights:

Stable

One component

Ready-to-use

Effectively inhibits peroxidase activity without interfering with antigen-antibody interactions

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.